Novus Biologicals
Manufacturer Code:NBP287578
Catalog # NB035200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human HMBOX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ21616 HNF1LA homeobox containing 1 homeobox-containing protein 1 homeobox-containing protein PBHNF PBHNF; Homeobox telomere-binding protein 1; Homeobox-containing protein 1; homeobox-containing protein PBHNF; Telomere-associated homeobox-containing protein 1
Gene Aliases: HMBOX1; HNF1LA; HOT1; PBHNF; TAH1
UniProt ID: (Human) Q6NT76
Entrez Gene ID: (Human) 79618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.