Proteintech
Manufacturer Code:161231AP150UL
Catalog # 50-173-0732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HMBOX1 Fusion Protein Ag9081 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HMBOX1 HNF1LA homeobox containing 1 Homeobox containing protein 1 PBHNF; homeobox containing 1; Homeobox telomere-binding protein 1; Homeobox-containing protein 1; homeobox-containing protein PBHNF; Telomere-associated homeobox-containing protein 1
Gene Aliases: AI451877; AI604847; F830020C16Rik; HMBOX1; HNF1LA; HOT1; PBHNF; RGD1306787; TAH1
UniProt ID: (Human) Q6NT76, (Mouse) Q8BJA3
Entrez Gene ID: (Human) 79618, (Rat) 305968, (Mouse) 219150
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.