Proteintech
Manufacturer Code:143361AP150UL
Catalog # 50-172-9374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HLX Fusion Protein Ag5627 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H2.0 like homeobox H2.0 like homeobox protein HB24 HLX HLX1 Homeobox protein HB24 Homeobox protein HLX1; H2.0-like homeo box 1; H2.0-like homeo box-1; H2.0-like homeobox 1; H2.0-like homeobox protein; Homeobox protein HB24; Homeobox protein HLX1
Gene Aliases: HB24; HLX; HLX1
UniProt ID: (Human) Q14774, (Mouse) Q61670, (Rat) A0JPN1
Entrez Gene ID: (Human) 3142, (Mouse) 15284, (Rat) 364069
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.