Invitrogen
Manufacturer Code:MA538133
Catalog # PIMA538133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1982 |
Immunogen | A synthesized peptide derived from human HLTF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AF010600 BC057116 DNA-binding protein/plasminogen activator inhibitor 1 regulator DNA-binding protein/plasminogen activator inhibitor-1 regulator helicase like transcription factor helicase-like transcription factor HIP116 HIP116A HLTF HLTF1 mem P113 RING finger protein 80 RING-type E3 ubiquitin transferase HLTF RNF80 Smarca3 Snf2l3 SNF2-like 3 Sucrose nonfermenting protein 2-like 3 sucrose nonfermenting-like 3 SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily a member 3 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3 TNF-response element-binding protein ZBU1; DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor-1 regulator; Helicase-like transcription factor; HIP116; P113; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; TNF-response element-binding protein
Gene Aliases: AF010600; BC057116; HIP116; HIP116A; HLTF; HLTF1; P113; RNF80; SMARCA3; SNF2L3; ZBU1
UniProt ID: (Human) Q14527, (Mouse) Q6PCN7
Entrez Gene ID: (Human) 6596, (Mouse) 20585, (Rat) 295568
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.