Novus Biologicals
Manufacturer Code:NB1001041
Catalog # NB1001041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KQAKINEIRTLIDL corresponding to C-Terminus according to NP_003062.2 NP_620636.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor 1 regulator EC 3.6.1 EC 3.6.4.- EC 6.3.2.- helicase-like transcription factor HIP116ARING finger protein 80 HIP116matrix associated actin dependent regulator of chromatin RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3 subfamily a member 3 Sucrose nonfermenting protein 2-like 3 ZBU1; DNA-binding protein/plasminogen activator inhibitor-1 regulator; Helicase-like transcription factor; HIP116; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3
Gene Aliases: HIP116; HIP116A; HLTF; HLTF1; RNF80; SMARCA3; SNF2L3; ZBU1
UniProt ID: (Human) Q14527
Entrez Gene ID: (Human) 6596
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.