Novus Biologicals
Manufacturer Code:NBP267610
Catalog # NBP267610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SC06-78 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: FLJ51114 histocompatibility antigen HLA-DR alpha HLA class II histocompatibility antigen DR alpha chain HLA-DRA1 major histocompatibility complex class II DR alpha MHC cell surface glycoprotein MHC class II antigen DRA MLRW; histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; MHC cell surface glycoprotein; MHC class II antigen DRA
Gene Aliases: HLA-DRA; HLA-DRA1; MLRW
UniProt ID: (Human) P01903
Entrez Gene ID: (Human) 3122
Molecular Function: defense/immunity protein major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.