EMD Millipore
Manufacturer Code:MABF2169100UG
Catalog # MABF2169100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H84 |
Immunogen | KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal region of Human MHC class I antigen G. The immunogen sequence is conserved in all isoforms. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HLA class I histocompatibility antigen alpha chain GHLA G antigenMHC class I antigen G
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.