Proteintech
Manufacturer Code:217021AP150UL
Catalog # 50-173-3554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HLA-DRB5 Fusion Protein Ag16307 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DR beta 2 DR beta 5 Dw2 HLA DRB HLA DRB5 HLA-DRB5 MHC class II antigen DRB5; DR beta-5; DR-9; DR2-beta-2; DR9; Dw2; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; HLA-DRB5; human leucocyte antigen DRB5; leukocyte antigen class II; MHC class I antigen; MHC class II antigen DRB1*9; MHC class II antigen DRB5; MHC class II HLA beta chain; MHC HLA-DR-beta cell surface glycoprotein; MHC HLA-DR-beta chain
Gene Aliases: HLA-DRB; HLA-DRB5
UniProt ID: (Human) Q30154
Entrez Gene ID: (Human) 3127
Molecular Function: defense/immunity protein major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.