Invitrogen
Manufacturer Code:MA518018
Catalog # PIMA518018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | YE2/36 HLK |
Immunogen | EHR-B Ramos cells |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen DR alpha chain MHC cell surface glycoprotein MHC class II antigen DRA histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; MHC cell surface glycoprotein; MHC class II antigen DRA
Gene Aliases: HLA-DRA; HLA-DRA1; MLRW
UniProt ID: (Human) P01903
Entrez Gene ID: (Human) 3122
Molecular Function: defense/immunity protein major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.