Invitrogen
Manufacturer Code:12995241
Catalog # 50-113-12
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | L243 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; MHC cell surface glycoprotein; MHC class II antigen DRA; XXbac-BPG161M6.1 DRB1 DRw10 FLJ75017 FLJ76359 HLA-DR1B HLA-DRB HLA-DRB1* SS1
Gene Aliases: HLA-DRA; HLA-DRA1; MLRW
UniProt ID: (Human) P01903
Entrez Gene ID: (Human) 3122
Molecular Function: defense/immunity protein major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.