Novus Biologicals
Manufacturer Code:NB1002700AF488
Catalog # NB1002EAFC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TAL 4.1 |
Immunogen | Whole cells from the B line WT46 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD CELIAC1DQ alpha 1 chain DC-1 alpha chain DQ-A1 FLJ27088 FLJ27328 GSE HLA class II histocompatibility antigen DQ(W3) alpha chain HLA-DCA HLA-DQA leucocyte antigen DQA1 leukocyte antigen alpha chain major histocompatibility complex class II DQ alpha 1 MGC149527 MHC class II antigen MHC class II DQA1 MHC class II HLA-D alpha glycoprotein MHC class II HLA-DQ-alpha-1 MHC class II surface glycoprotein MHC HLA-DQ alpha; DC-1 alpha chain; DC-alpha; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA-DCA; leucocyte antigen DQA1; leukocyte antigen alpha chain; MHC class II antigen; MHC class II DQA1; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DQ-alpha-1; MHC class II surface glycoprotein; MHC HLA-DQ alpha
Gene Aliases: CD; CELIAC1; DQ-A1; GSE; HLA-DQA; HLA-DQA1
UniProt ID: (Human) P01909
Entrez Gene ID: (Human) 3117
Molecular Function: defense/immunity protein major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.