Abnova
Manufacturer Code:H00007917B01P
Catalog # 89-016-938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | BAT3 (NP_542433.1 1 a.a. ~ 1126 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human BAT3 protein.
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 6; BAG-6; BAG6; BCL2-associated athanogene 6; D6S52E; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; Protein G3; Protein Scythe; scythe
Gene Aliases: BAG-6; BAG6; BAT3; D6S52E; G3
UniProt ID: (Human) P46379
Entrez Gene ID: (Human) 7917
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.