Novus Biologicals
Manufacturer Code:NBP261871
Catalog # NBP2-61871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G7B10 |
Immunogen | Purified recombinant fragment of human HLA-B (AA: 241-362) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: ankylosing spondylitis AS b&primeDT Bw-22 Bw-4 Bw-41 Bw-44 Bw-45 Bw-46 Bw-47 Bw-48 Bw-50 Bw-52 Bw-53 Bw-54 Bw-55 Bw-56 Bw-57 Bw-58 Bw-60 HLA class I histocompatibility antigen B alpha chain HLA class I histocompatibility antigen B-12 alpha chain HLA class I histocompatibility antigen B-21 alpha chain HLA class I histocompatibility antigen B-5 alpha chain HLA class I histocompatibility antigen B-7 alpha chain HLAB HLA-B27 HLA-B73 leukocyte antigen class I-B lymphocyte antigen major histocompatibility complex class I B MGC111087 MHC class I antigen B&ast13 MHC class I antigen B&ast14 MHC class I antigen B&ast15 MHC class I antigen B&ast18 MHC class I antigen B&ast27 MHC class I antigen B&ast35 MHC class I antigen B&ast37 MHC class I antigen B&ast38 MHC class I antigen B&ast39 MHC class I antigen B&ast40 MHC class I antigen B&ast41 MHC class I antigen B&ast42 MHC class I antigen B&ast44 MHC class I antigen B&ast45 MHC class I antigen B&ast46 MHC class I antigen B&ast47 MHC class I anti; class I MHC, HLA-B antigen; epididymis secretory protein Li 83; histocompatibility antigen; HLA -B; HLA B-40011; HLA B44; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLA-B; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B35; HLA-B35 antigen; HLA-Bw50 antigen; Human leukocyte antigen B; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27-HS; MHC HLA-B27d; MHC HLA-B39N; truncated class I antigen; truncated MHC class I antigen
Gene Aliases: AS; B-4901; B-5001; Bw-47; Bw-50; HEL-S-83; HLA-B; HLA-B*45ZJ; HLA-B-3506; HLA-B-3905; HLA-B-5502; HLA-B-5602; HLA-B15; HLA-B39; HLA-B49; HLA-B50; HLA-B55; HLA-B59; HLA-B61; HLA-Cw; HLA-DRB1; HLAB; SPDA1
UniProt ID: (Human) P79523
Entrez Gene ID: (Human) 3106
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.