Proteintech
Manufacturer Code:258741AP150UL
Catalog # 50-173-5111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HKDC1 Fusion Protein Ag23045 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hexokinase domain containing 1 HKDC1 Putative hexokinase HKDC1; Hexokinase domain-containing protein 1; Hexokinase HKDC1; putative hexokinase HKDC1
Gene Aliases: BC016235; HKDC1
UniProt ID: (Human) Q2TB90, (Mouse) Q91W97
Entrez Gene ID: (Human) 80201, (Mouse) 216019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.