Abnova
Manufacturer Code:MAB12977
Catalog # 89-934-253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F5 |
Immunogen | Recombinant protein corresponding to full length human HK3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full length recombinant human HK3.
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; hexokinase 3 (white cell); Hexokinase type III; Hexokinase-3; Hexokinase-C; HK III; HKIII; HXK3
Gene Aliases: HK3; HKIII; HXK3
UniProt ID: (Human) P52790
Entrez Gene ID: (Human) 3101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.