Invitrogen
Manufacturer Code:PA528666
Catalog # PIPA528666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 535 and 923 of Human Hexokinase III |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; hexokinase 3 (white cell); Hexokinase type III; Hexokinase-3; Hexokinase-C; HK III; HKIII HXK3 ATP:D-hexose 6-phosphotransferase HK III hexokinase type III hexokinase-3
Gene Aliases: HK3; HKIII; HXK3
UniProt ID: (Human) P52790
Entrez Gene ID: (Human) 3101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.