Invitrogen
Manufacturer Code:PA515442
Catalog # PIPA515442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HK3 (Hexokinase III) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; hexokinase 3 (white cell); Hexokinase type III; Hexokinase type III HK III; Hexokinase-3; Hexokinase-C; HK III
Gene Aliases: HK3; HKIII; HXK3
UniProt ID: (Human) P52790
Entrez Gene ID: (Human) 3101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.