Invitrogen
Manufacturer Code:MA514849
Catalog # PIMA514849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | H.738.7 |
Immunogen | Synthetic peptide corresponding to the sequence of human |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Hexokinase type II; Hexokinase-2; hexokinase-2, muscle; Hexokinase-B; HK II; HK II OTTHUMP00000202738 hexokinase type II hexokinase-2 hexokinase-2 muscle muscle form hexokinase; Muscle form hexokinase
Gene Aliases: AI642394; HK2; HKII; HXK2
UniProt ID: (Human) P52789, (Mouse) O08528, (Rat) P27881
Entrez Gene ID: (Human) 3099, (Mouse) 15277, (Rat) 25059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.