Invitrogen
Manufacturer Code:PA579364
Catalog # PIPA579364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human Hexokinase II. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hexokinase type II; Hexokinase-2; Hexokinase-2 Hexokinase type II HK II Muscle form hexokinase HK2; hexokinase-2, muscle; Hexokinase-B; HK II; Muscle form hexokinase
Gene Aliases: AI642394; HK2; HKII; HXK2
UniProt ID: (Human) P52789, (Mouse) O08528, (Rat) P27881
Entrez Gene ID: (Human) 3099, (Mouse) 15277, (Rat) 25059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.