Invitrogen
Manufacturer Code:PA529326
Catalog # PIPA529326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 53 and 342 of Human Hexokinase II |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hexokinase II; Hexokinase type II; Hexokinase-2; hexokinase-2, muscle; Hexokinase-B; HK II; HKII HXK2 HK II hexokinase type II hexokinase-2 hexokinase-2 muscle muscle form hexokinase; Muscle form hexokinase
Gene Aliases: AI642394; HK2; HKII; HXK2
UniProt ID: (Human) P52789, (Horse) A2PYL6, (Mouse) O08528, (Rat) P27881
Entrez Gene ID: (Human) 3099, (Horse) 100009677, (Mouse) 15277, (Rat) 25059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.