Invitrogen
Manufacturer Code:PA515443
Catalog # PIPA515443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 91-121 amino acids from the N-terminal region of human HK2 (Hexokinase II) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hexokinase type II; Hexokinase type II HK II Muscle form hexokinase; Hexokinase-2; hexokinase-2, muscle; Hexokinase-B; HK II; Muscle form hexokinase
Gene Aliases: HK2; HKII; HXK2
UniProt ID: (Human) P52789
Entrez Gene ID: (Human) 3099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.