Abnova
Manufacturer Code:MAB10683
Catalog # 89-108-883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A10 |
Immunogen | Recombinant protein corresponding to human HK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant HK1.
For Research Use Only
Protein Aliases: Brain form hexokinase; glycolytic enzyme; hexokinase I; hexokinase IR; Hexokinase type I; Hexokinase-1; Hexokinase-A; HK I; HK1-ta; HK1-tb; HK1-tc; HKI; HXK1
Gene Aliases: hexokinase; HK; HK1; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1
UniProt ID: (Human) P19367
Entrez Gene ID: (Human) 3098
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.