Novus Biologicals
Manufacturer Code:NBP252482
Catalog # NBP252482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E9B11 |
Immunogen | Synthesized peptide of human HH3 (AA: ARTKQTAR(AcK)STG-C). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H3 histone family member T H3/gH3.4 H3/t H3FTMGC126888 H3T H3tMGC126886 histone 3 H3 histone cluster 3 H3 histone H3.1t; H3 histone family, member T; H3/g; H3/t; histone 3, H3; Histone H3.1t; Histone H3.4
Gene Aliases: H3-4; H3.4; H3/g; H3FT; H3t; HIST3H3
UniProt ID: (Human) Q16695
Entrez Gene ID: (Human) 8290
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.