Novus Biologicals
Manufacturer Code:NBP298035FR
Catalog # NB051405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human HISPPD1 fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diphosphoinositol pentakisphosphate kinase 2; diphosphoinositol pentakisphosphate kinase 2FLJ21506 EC 2.7.4.21 EC 2.7.4.24 HISPPD1histidine acid phosphatase domain containing 1 Histidine acid phosphatase domain-containing protein 1 hsVIP2 inositol heptaphosphate kinase 2 inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2 InsP6 and PP-IP5 kinase 2 KIAA0433FLJ23463 VIP1 homolog 2 VIP2IP7K2; Histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; VIP1 homolog 2
Gene Aliases: CFAP160; HISPPD1; IP7K2; KIAA0433; PPIP5K2; VIP2
UniProt ID: (Human) O43314
Entrez Gene ID: (Human) 23262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.