Invitrogen
Manufacturer Code:PA540567
Catalog # PIPA540567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human HIPK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hHIPk2; Homeodomain-interacting protein kinase 2; Nuclear body-associated kinase 1; Sialophorin tail-associated nuclear serine/threonine-protein kinase
Gene Aliases: 1110014O20Rik; B230339E18Rik; cb1009; HIPK2; Nbak1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Stank; wu:fk56d12
UniProt ID: (Human) Q9H2X6, (Mouse) Q9QZR5
Entrez Gene ID: (Dog) 482772, (Bovine) 510158, (Human) 28996, (Rabbit) 100359086, (Horse) 100064748, (Rat) 362342, (Guinea pig) 100729474, (Mouse) 15258, (Zebrafish) 557551
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.