OriGene
Manufacturer Code:CF807258
Catalog # 50-166-8178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 439-697 of human HIPK1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: homeodomain interacting protein kinase 1-like protein; Homeodomain-interacting protein kinase 1; Myak Nbak2; nuclear body associated kinase 2b; Nuclear body-associated kinase 2
Gene Aliases: HIPK1; KIAA0630; MYAK; NBAK2
UniProt ID: (Human) Q86Z02
Entrez Gene ID: (Human) 204851
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.