Invitrogen
Manufacturer Code:PA528799
Catalog # PIPA528799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1147 and 1210 of Human HIPK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeodomain interacting protein kinase 1-like protein; Homeodomain-interacting protein kinase 1; Myak Nbak2 homeodomain interacting protein kinase 1-like protein homeodomain-interacting protein kinase 1 nuclear body associated kinase 2b nuclear body-associated kinase 2; nuclear body associated kinase 2b; Nuclear body-associated kinase 2; Protein kinase Myak
Gene Aliases: 1110062K04Rik; HIPK1; KIAA0630; MYAK; NBAK2
UniProt ID: (Human) Q86Z02, (Mouse) O88904
Entrez Gene ID: (Human) 204851, (Mouse) 15257
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.