Abnova
Manufacturer Code:H00009026M01A
Catalog # 89-105-995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E10 |
Immunogen | HIP1R (NP_003950969 a.a. &bsim 1066 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HIP1R.
For Research Use Only
Protein Aliases: FLJ14000; FLJ27022; HIP-12; HIP1-related protein; HIP12; HIP3; huntingtin interacting protein 12; Huntingtin-interacting protein 1-related protein; Huntingtin-interacting protein 12; ILWEQ; KIAA0655; MGC47513
Gene Aliases: HIP12; HIP1R; HIP3; ILWEQ; KIAA0655
UniProt ID: (Human) O75146
Entrez Gene ID: (Human) 9026
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.