Invitrogen
Manufacturer Code:MA116750
Catalog # AMPA6750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E1 |
Immunogen | Human HIP1r-TALIN Homology Region (57 kDa). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HIP-12; HIP1-related protein; HIP12 HIP3 ILWEQ HIP-12 HIP1-related protein huntingtin interacting protein 12 huntingtin-interacting protein 1-related protein huntingtin-interacting protein 12; huntingtin interacting protein 12; Huntingtin-interacting protein 1-related protein; Huntingtin-interacting protein 12
Gene Aliases: AA410023; HIP12; HIP1R; HIP3; ILWEQ; KIAA0655; mKIAA0655
UniProt ID: (Human) O75146, (Mouse) Q9JKY5
Entrez Gene ID: (Human) 9026, (Mouse) 29816, (Rat) 81917
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.