Proteintech
Manufacturer Code:154651AP150UL
Catalog # 50-173-0250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HIP14 ZDHHC17 Fusion Protein Ag7763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyltransferase ZDHHC17; DHHC 17 HIP 14 HIP 3 HIP14 HIP3 HSPC294 Huntingtin yeast partner H HYPH KIAA0946 Palmitoyltransferase ZDHHC17 ZDHHC17; DHHC domain-containing cysteine-rich protein 17; DHHC-17; DHHC17; HIP-14; HIP-3; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; Huntingtin yeast partner H; Huntingtin-interacting protein 14; Huntingtin-interacting protein 3; Huntingtin-interacting protein H; membrane-associated DHHC17 zinc finger protein; Palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; Putative NF-kappa-B-activating protein 205; Zinc finger DHHC domain-containing protein 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17
Gene Aliases: A230053P19Rik; BB187739; D130071N24Rik; HIP14; HIP3; HSPC294; HYPH; KIAA0946; ZDHHC17
UniProt ID: (Human) Q8IUH5, (Mouse) Q80TN5, (Rat) E9PTT0
Entrez Gene ID: (Human) 23390, (Mouse) 320150, (Rat) 366889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.