Invitrogen
Manufacturer Code:PA532600
Catalog # PIPA532600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from internal region of human ST13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aging-associated protein 2; heat shock 70kD protein binding protein; Hip; Hsc70-interacting protein; Hsp70-interacting protein; Hsp70-interacting protein aging-associated protein 2 heat shock 70kD protein binding protein hsc70-interacting protein progesterone receptor-associated p48 protein putative tumor suppressor ST13 AAG2 FAM10A1 HIP SNC6 renal carcinoma antigen NY-REN-33 suppression of tumorigenicity 13 protein Protein FAM10A1; Progesterone receptor-associated p48 protein; Protein FAM10A1; Putative tumor suppressor ST13; Renal carcinoma antigen NY-REN-33; Suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m
Gene Aliases: AAG2; FAM10A1; FAM10A4; HIP; HOP; HSPABP; HSPABP1; P48; PRO0786; SNC6; ST13
UniProt ID: (Human) P50502
Entrez Gene ID: (Human) 6767
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.