Invitrogen
Manufacturer Code:MA524736
Catalog # PIMA524736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1500CT836.13.93 |
Immunogen | human HINT1 recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 17 kDa inhibitor of protein kinase C; Adenosine 5'-monophosphoramidase; Histidine triad nucleotide-binding protein 1; Histidine triad nucleotide-binding protein 1 3--- Adenosine 5&prime-monophosphoramidase Protein kinase C inhibitor 1 Protein kinase C-interacting protein 1 PKCI-1 HINT1 HINT PKCI1 PRKCNH1; PKC inhibitor/ interacting protein; PKCI-1; Protein kinase C inhibitor 1; protein kinase C, iota; Protein kinase C-interacting protein 1
Gene Aliases: AA673479; HINT; HINT1; Ipk1; NMAN; Pkci; PKCI-1; PKCI1; Prkci; PRKCNH1
UniProt ID: (Human) P49773, (Mouse) P70349, (Rat) P62959
Entrez Gene ID: (Human) 3094, (Mouse) 15254, (Rat) 690660
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.