Invitrogen
Manufacturer Code:PA5115357
Catalog # PIPA5115357
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HINT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17 kDa inhibitor of protein kinase C; 17 kDa inhibitor of protein kinase C AA673479 Adenosine 5'-monophosphoramidase FLJ30414 FLJ32340 HINT HINT1 histidine triad nucleotide binding protein 1 histidine triad nucleotide-binding protein 1 Ipk1 NMAN PKC inhibitor/ interacting protein Pkci Pkci1 PKCI-1 Prkci PRKCNH1 Protein kinase C inhibitor 1 protein kinase C iota protein kinase C-interacting protein 1; Adenosine 5'-monophosphoramidase; Histidine triad nucleotide-binding protein 1; PKC inhibitor/ interacting protein; PKCI-1; Protein kinase C inhibitor 1; protein kinase C, iota; Protein kinase C-interacting protein 1
Gene Aliases: AA673479; HINT; HINT1; Ipk1; NMAN; Pkci; PKCI-1; PKCI1; Prkci; PRKCNH1
UniProt ID: (Human) P49773, (Mouse) P70349, (Rat) P62959
Entrez Gene ID: (Human) 3094, (Mouse) 15254, (Rat) 690660
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.