Novus Biologicals
Manufacturer Code:NBP183257
Catalog # NBP183257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IPKKHISQISVAEDDDESLLGHLMIVGKKCAADLGLNKGYRMVVNEGSDGGQSVYHVHLHVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine 5'-monophosphoramidase; Adenosine 5'-monophosphoramidase EC 3.- HINT histidine triad nucleotide binding protein 1 histidine triad nucleotide-binding protein histidine triad nucleotide-binding protein 1 PKCI1 PKCI-1FLJ30414 PRKCNH1FLJ32340 Protein kinase C inhibitor 1 Protein kinase C-interacting protein 1; Histidine triad nucleotide-binding protein 1; PKCI-1; Protein kinase C inhibitor 1; Protein kinase C-interacting protein 1
Gene Aliases: HINT; HINT1; NMAN; PKCI-1; PKCI1; PRKCNH1
UniProt ID: (Human) P49773
Entrez Gene ID: (Human) 3094
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.