Novus Biologicals
Manufacturer Code:NBP30325320UL
Catalog # NB092946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-126 of human HINT1 (NP_005331.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine 5'-monophosphoramidase; Adenosine 5'-monophosphoramidase EC 3.- HINT histidine triad nucleotide binding protein 1 histidine triad nucleotide-binding protein histidine triad nucleotide-binding protein 1 PKCI1 PKCI-1FLJ30414 PRKCNH1FLJ32340 Protein kinase C inhibitor 1 Protein kinase C-interacting protein 1; Histidine triad nucleotide-binding protein 1; PKCI-1; Protein kinase C inhibitor 1; Protein kinase C-interacting protein 1
Gene Aliases: HINT; HINT1; NMAN; PKCI-1; PKCI1; PRKCNH1
UniProt ID: (Human) P49773
Entrez Gene ID: (Human) 3094
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.