Abnova
Manufacturer Code:PAB16555
Catalog # 89-118-424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region 17 amino acids of human HINT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HINT1.
For Research Use Only
Protein Aliases: Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Histidine triad nucleotide-binding protein 1; PKCI-1; PRKCNH1; Protein kinase C inhibitor 1; Protein kinase C-interacting protein 1
Gene Aliases: HINT; HINT1; NMAN; PKCI-1; PKCI1; PRKCNH1
UniProt ID: (Human) P49773
Entrez Gene ID: (Human) 3094
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.