Proteintech
Manufacturer Code:106461AP150UL
Catalog # 50-172-6476
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HIF1AN Fusion Protein Ag0997 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Factor inhibiting HIF 1 FIH 1 FIH1 HIF1AN; Factor inhibiting HIF-1; factor inhibiting HIF1; FIH-1; hypoxia inducible factor 1, alpha subunit inhibitor; Hypoxia-inducible factor 1-alpha inhibitor; Hypoxia-inducible factor asparagine hydroxylase; peptide-aspartate beta-dioxygenase
Gene Aliases: 2310046M24Rik; A830014H24Rik; FIH; FIH1; HIF1AN
UniProt ID: (Human) Q9NWT6, (Mouse) Q3U3G4
Entrez Gene ID: (Human) 55662, (Rat) 309434, (Mouse) 319594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.