Invitrogen
Manufacturer Code:MA516019
Catalog # PIMA516019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ESEE122 |
Immunogen | Human HIF-1 alpha amino acids 329-530. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARNT interacting protein; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF-1 alpha; HIF-1-alpha; HIF-1alpha HIF1 HIF1-ALPHA MOP1 PASD8 bHLHe78 ARNT interacting protein ARNT-interacting protein HIF-1-alpha PAS domain-containing protein 8 basic-helix-loop-helix-PAS protein MOP1 class E basic helix-loop-helix protein 78 hypoxia-inducible factor 1 alpha isoform I.3 hypoxia-inducible factor 1 alpha subunit (basic helix-loop-helix transcription factor) hypoxia-inducible factor 1-alpha member of PAS protein 1 member of PAS superfamily 1; HIF1 alpha; HIF1-alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); Hypoxia-inducible factor 1-alpha; hypoxia-inducible factor1alpha; Member of PAS protein 1; member of PAS superfamily 1; PAS domain-containing protein 8
Gene Aliases: AA959795; BHLHE78; HIF-1-alpha; HIF-1A; HIF-1alpha; HIF1; HIF1-ALPHA; HIF1A; HIF1alpha; MOP1; PASD8
UniProt ID: (Human) Q16665, (Mouse) Q61221, (Rat) O35800
Entrez Gene ID: (Bovine) 281814, (Human) 3091, (Dog) 480348, (Mouse) 15251, (Rat) 29560
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.