Invitrogen
Manufacturer Code:PA585494
Catalog # PIPA585494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human HIF1 alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARNT interacting protein antibody; ARNT-interacting protein antibody; basic-helix-loop-helix-PAS protein MOP1 antibody; bHLHe78 antibody; class E basic helix-loop-helix protein 78 antibody; HIF-1-alpha antibody; HIF1 antibody; HIF1-ALPHA antibody; HIF1A antibody; hypoxia inducible factor 1 alpha subunit (basic helix-loop-helix transcription factor) antibody; hypoxia-inducible factor 1 alpha isoform I.3 antibody; hypoxia-inducible factor 1 alpha subunit (basic helix-loop-helix transcription factor) antibody; hypoxia-inducible factor 1-alpha antibody; member of PAS protein 1 antibody; member of PAS superfamily 1 antibody; MOP1 antibody; PAS domain-containing protein 8 antibody; PASD8 antibody; ARNT interacting protein; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF-1 alpha; HIF-1-alpha; HIF-1alpha antibody; HIF1 alpha; HIF1-alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); Hypoxia-inducible factor 1-alpha; hypoxia-inducible factor1alpha; Member of PAS protein 1; member of PAS superfamily 1; PAS domain-containing protein 8
Gene Aliases: AA959795; BHLHE78; HIF-1-alpha; HIF-1A; HIF-1alpha; HIF1; HIF1-ALPHA; HIF1A; HIF1alpha; MOP1; PASD8
UniProt ID: (Human) Q16665, (Mouse) Q61221, (Rat) O35800
Entrez Gene ID: (Bovine) 281814, (Human) 3091, (Mouse) 15251, (Rat) 29560
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.