Invitrogen
Manufacturer Code:MA516021
Catalog # PIMA516021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ep190b |
Immunogen | Human HIF-2 alpha amino acids 535-631. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4 HIF2A HLF MOP2 PASD2 bHLHe73 EPAS-1 HIF-1-alpha-like factor HIF-1alpha-like factor HIF-2-alpha HIF2-alpha PAS domain-containing protein 2 basic-helix-loop-helix-PAS protein MOP2 class E basic helix-loop-helix protein 73 endothelial PAS domain-containing protein 1 hypoxia-inducible factor 2 alpha hypoxia-inducible factor 2-alpha member of PAS protein 2; endothelial PAS domain protein 1; Endothelial PAS domain-containing protein 1; EPAS-1; Hif like protein; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF-2 alpha; HIF-2-alpha; HIF-related factor; HIF1 alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF2-alpha; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; Hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; PAS domain-containing protein 2
Gene Aliases: BHLHE73; ECYT4; EPAS1; HIF-2alpha; HIF2A; HLF; HRF; MOP2; PASD2
UniProt ID: (Human) Q99814, (Mouse) P97481, (Rat) Q9JHS1
Entrez Gene ID: (Human) 2034, (Mouse) 13819, (Chinese hamster) 100771160, (Rat) 29452
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.