Invitrogen
Manufacturer Code:PA585190
Catalog # PIPA585190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human HIF2 alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4 antibody HIF2A antibody HLF antibody MOP2 antibody PASD2 antibody bHLHe73 antibody EPAS1 antibody hypoxia-inducible factor 2-alpha antibody HIF-2-alpha antibody hypoxia-inducible factor 2 alpha antibody member of PAS protein 2 antibody EPAS-1 antibody HIF-1alpha-like factor antibody PAS domain-containing protein 2 antibody class E basic helix-loop-helix protein 73 antibody HIF2-alpha antibody endothelial PAS domain-containing protein 1 antibody basic-helix-loop-helix-PAS protein MOP2 antibody HIF-1-alpha-like factor antibody endothelial PAS domain protein 1 antibody; endothelial PAS domain protein 1; Endothelial PAS domain-containing protein 1; EPAS-1; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF-2-alpha; hif-2a; HIF-2alpha; HIF2-alpha; HIF2A; HLF; hypoxia inducible factor 2 alpha; hypoxia-inducible factor 2 alpha; Hypoxia-inducible factor 2-alpha; Member of PAS protein 2; PAS domain-containing protein 2
Gene Aliases: BHLHE73; ECYT4; EPAS1; hif-2ab; HIF2A; HLF; MOP2; PASD2; wu:fq36f01
UniProt ID: (Human) Q99814
Entrez Gene ID: (Human) 2034, (Zebrafish) 555192
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.