Invitrogen
Manufacturer Code:MA516015
Catalog # PIMA516015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H1beta234 |
Immunogen | Fusion protein containing amino acids 496-789 of human HIF-1 beta. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARNT; ARNT protein; Aryl hydrocarbon receptor nuclear translocator; Aryl hydrocarbon receptor nuclear translocator 1; bHLHe2; Class E basic helix-loop-helix protein 2; Dioxin receptor, nuclear translocator; HIF-1 beta; HIF-1-beta; HIF-1-beta HIF-1beta HIF1-beta HIF1B HIF1BETA TANGO bHLHe2 class E basic helix-loop-helix protein 2 dioxin receptor nuclear translocator hypoxia-inducible factor 1 beta subunit; HIF1-beta; hypoxia-inducible factor 1 beta; hypoxia-inducible factor 1, beta subunit; Hypoxia-inducible factor 1-beta
Gene Aliases: ARNT; Arnt1; BHLHE2; D3Ertd557e; Drnt; ESTM42; HIF-1-beta; HIF-1beta; HIF1-beta; HIF1B; HIF1BETA; mKIAA4051; TANGO; W08714
UniProt ID: (Human) P27540, (Bovine) Q9BE97, (Rat) P41739, (Mouse) P53762
Entrez Gene ID: (Human) 405, (Bovine) 281010, (Sheep) 443285, (Rat) 25242, (Mouse) 11863
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.