R&D Systems
Manufacturer Code:MAB19351100
Catalog # MAB19351100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2443B |
Immunogen | <em>E. coli</em>-derived recombinant human HIF-1 alpha Arg575-Asn826 Accession # Q16665.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
Protein Aliases: AINT anti-HIF-1 alpha anti-HIF1A ARNT interacting protein ARNT-interacting protein Basic-helix-loop-helix-PAS protein MOP1 BHLHE78 Class E basic helix-loop-helix protein 78 HIF 1A HIF1 HIF1 alpha HIF-1 alpha HIF-1a HIF1A HIF-1alpha HIF-1-alpha HIF1-alpha hypoxia inducible factor 1 alpha subunit hypoxia inducible factor 1 subunit alpha hypoxia inducible factor 1 alpha subunit (basic helix-loop-helix transcription factor) hypoxia-inducible factor 1-alpha Member of PAS protein 1 member of PAS superfamily 1 MOP1 PAS domain-containing protein 8 PASD8; ARNT interacting protein; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF-1-alpha; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); Hypoxia-inducible factor 1-alpha; hypoxia-inducible factor1alpha; Member of PAS protein 1; member of PAS superfamily 1; PAS domain-containing protein 8
Gene Aliases: BHLHE78; HIF-1-alpha; HIF-1A; HIF-1alpha; HIF1; HIF1-ALPHA; HIF1A; MOP1; PASD8
UniProt ID: (Human) Q16665
Entrez Gene ID: (Human) 3091
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.