Invitrogen
Manufacturer Code:PA568597
Catalog # PIPA568597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human HIC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; Androgen receptor-associated protein of 55 kDa; ARA55 HIC-5 HIC5 TSC-5 androgen receptor coactivator 55 kDa protein androgen receptor coactivator ARA55 androgen receptor-associated protein of 55 kDa hydrogen peroxide-inducible clone 5 protein transforming growth factor beta-1-induced transcript 1 protein; Hic-5; Hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Aliases: ARA55; HIC-5; HIC5; TGFB1I1; TSC-5
UniProt ID: (Bovine) Q3MHZ4, (Human) O43294, (Mouse) Q62219, (Rat) Q99PD6
Entrez Gene ID: (Dog) 489932, (Rabbit) 100340706, (Goat) 100861032, (Bovine) 515834, (Human) 7041, (Horse) 100063161, (Mouse) 21804, (Rat) 84574, (Guinea pig) 100719547
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.