Abnova
Manufacturer Code:H00003090M01
Catalog # 89-022-484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E11 |
Immunogen | HIC1 (NP_006488 396 a.a. ~ 453 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HIC1.
For Research Use Only
Protein Aliases: Hic-1; Hypermethylated in cancer 1 protein; ZBTB29; Zinc finger and BTB domain-containing protein 29
Gene Aliases: hic-1; HIC1; ZBTB29; ZNF901
UniProt ID: (Human) Q14526
Entrez Gene ID: (Human) 3090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.