Proteintech
Manufacturer Code:249491AP150UL
Catalog # 50-173-4638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HIC1 Fusion Protein Ag21849 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hic 1 HIC1 hypermethylated in cancer 1 ZBTB29; Hic-1; Hypermethylated in cancer 1 protein; Zinc finger and BTB domain-containing protein 29
Gene Aliases: AA408311; HIC-1; HIC1; ZBTB29; ZNF901
UniProt ID: (Human) Q14526, (Mouse) Q9R1Y5
Entrez Gene ID: (Human) 3090, (Mouse) 15248
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.