OriGene
Manufacturer Code:CF501319
Catalog # 50-166-0342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C7 |
Immunogen | Full length human recombit protein of human HIBCH produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; 3-hydroxyisobutyryl-Coenzyme A hydrolase; HIB-CoA hydrolase; HIBYL-CoA-H; HIBYLCOAH; testicular tissue protein Li 86
Gene Aliases: HIBCH; HIBYLCOAH
UniProt ID: (Human) Q6NVY1, (Rat) Q5XIE6
Entrez Gene ID: (Dog) 607040, (Human) 26275, (Rat) 301384
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.