Proteintech
Manufacturer Code:146031AP150UL
Catalog # 50-172-9576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HIBCH Fusion Protein Ag6149 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; 3-hydroxyisobutyryl-Coenzyme A hydrolase; HIB CoA hydrolase HIBCH HIBYL COA H; HIB-CoA hydrolase; HIBYL-COA-H; testicular tissue protein Li 86
Gene Aliases: 2610509I15Rik; AI648812; HIBCH; HIBYLCOAH
UniProt ID: (Human) Q6NVY1, (Rat) Q5XIE6, (Mouse) Q8QZS1
Entrez Gene ID: (Human) 26275, (Rat) 301384, (Mouse) 227095
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.