Invitrogen
Manufacturer Code:PA570828
Catalog # PIPA570828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human HHIPL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hedgehog interacting protein-like 2; HHIP-like 2; HHIP-like protein 2; KIAA1822L HHIP-like protein 2 hedgehog interacting protein-like 2; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa
Gene Aliases: HHIP3; HHIPL2; KIAA1822L; UNQ841/PRO1779
UniProt ID: (Human) Q6UWX4
Entrez Gene ID: (Dog) 478998, (Human) 79802, (Horse) 100055380, (Pig) 100517774
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.