Invitrogen
Manufacturer Code:PA563765
Catalog # PIPA563765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HHIPL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hedgehog interacting protein-like 2; HHIP-like 2; HHIP-like protein 2; KIAA1822L HHIP-like protein 2 hedgehog interacting protein-like 2
Gene Aliases: HHIP3; HHIPL2; KIAA1822L; UNQ841/PRO1779
UniProt ID: (Human) Q6UWX4
Entrez Gene ID: (Human) 79802
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.